missing translation for 'onlineSavingsMsg'
Learn More

2-Formylbenzenesulfonic acid, sodium salt, 90%, tech.

Product Code. 10043863 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
2.5kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10043863 2.5 kg Plastic bottle 2.5kg
10175130 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10043863 Supplier Thermo Scientific Acros Supplier No. 219320025
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Añadir a la cesta
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 1008-72-6 | C7H5NaO4S | 208.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 1008-72-6
Molecular Formula C7H5NaO4S
Molecular Weight (g/mol) 208.16
MDL Number MFCD00007478
InChI Key ADPUQRRLAAPXGT-UHFFFAOYSA-M
Synonym sodium 2-formylbenzenesulfonate, 2-formylbenzenesulfonic acid sodium salt, sodium o-formylbenzenesulfonate, benzenesulfonic acid, 2-formyl-, sodium salt, unii-lh8xh59068, 2-sulfobenzaldehyde sodium salt, sodium benzaldehyde-2-sulfonate, benzaldehyde-2-sulfonic acid sodium salt, o-formylbenzenesulfonic acid sodium salt, sodium 2-formylbenzene-1-sulfonate
PubChem CID 3794540
IUPAC Name sodium;2-formylbenzenesulfonate
SMILES [Na+].[O-]S(=O)(=O)C1=CC=CC=C1C=O

Especificaciones

CAS Min % 88.0
CAS Max % 100.0
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Plastic bottle
Linear Formula HCOC6H4SO3Na
Quantity 2.5 kg
Beilstein 11, 324
Formula Weight 208.17
Percent Purity 90%
Grade Technical
Physical Form Powder
Chemical Name or Material 2-Formylbenzenesulfonic acid sodium salt, tech., 90%
Mostrar más Mostrar menos

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.