missing translation for 'onlineSavingsMsg'
Learn More

2-Furaldehyde, ACS, 98% min

Product Code. 11357448
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357448 250 g 250g
11367448 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357448 Supplier Thermo Scientific Alfa Aesar Supplier No. 031305.30
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 98-01-1 | C5H4O2 | 96.09 g/mol

It is primarily used in the production of furan derivatives such as furan and furfuryl alcohol. Another major application of furfural is as extraction solvent in refineries. It is also used to accelerate vulcanization, as an ingredient of phenolic resins (Durite) and as a flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-01-1
Molecular Formula C5H4O2
Molecular Weight (g/mol) 96.09
MDL Number MFCD00003229
InChI Key HYBBIBNJHNGZAN-UHFFFAOYSA-N
Synonym furfural, 2-furaldehyde, 2-furancarboxaldehyde, furaldehyde, 2-formylfuran, furfuraldehyde, fural, 2-furanaldehyde, 2-furancarbonal, 2-furfural
PubChem CID 7362
ChEBI CHEBI:34768
IUPAC Name furan-2-carbaldehyde
SMILES O=CC1=CC=CO1

Specifications

Melting Point -38°C
Color Yellow to Brown
Density 1.159 g/mL
Boiling Point 162°C
Flash Point 60°C (140°F)
Odor Almond-like
Refractive Index 1.5261
Quantity 250 g
UN Number UN1199
Beilstein 105755
Sensitivity Air Sensitive
Merck Index 14,4304
Solubility Information It is soluble in water.
Formula Weight 96.09
Percent Purity ≤98%
Grade ACS
Physical Form Liquid
Chemical Name or Material 2-Furaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.