missing translation for 'onlineSavingsMsg'
Learn More

2-Furancarboxylic Acid Methyl Ester, TRC

Product Code. 30408336
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30408336 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30408336 Supplier Toronto Research Chemicals Supplier No. TRCF86376510G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl Furan-2-carboxylate
CAS 611-13-2
Molecular Formula C6 H6 O3
Quantity 10 g
Synonym AH 22254X, Fluticasone furoate related, Methyl 2-furoate
InChI Formula InChI=1S/C6H6O3/c1-8-6(7)5-3-2-4-9-5/h2-4H,1H3
SMILES COC(=O)c1occc1
IUPAC Name methyl furan-2-carboxylate
Molecular Weight (g/mol) 126.11
Formula Weight 126.0317
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.