missing translation for 'onlineSavingsMsg'
Learn More

2-Furylacetone, 99%

Product Code. 11499697
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499697 5 g 5g
11409707 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11499697 Supplier Thermo Scientific Alfa Aesar Supplier No. B22171.06
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6975-60-6 | C7H8O2 | 124.139 g/mol

Suitable for fragrance applications.2-Furylacetone is used as the reactant in the synthesis and transformations of furan derivatives

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Suitable for fragrance applications.2-Furylacetone is used as the reactant in the synthesis and transformations of furan derivatives

Solubility
Soluble in alcohol, triacetin.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6975-60-6
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.139
MDL Number MFCD02683084
InChI Key IQOJTGSBENZIOL-UHFFFAOYSA-N
Synonym 2-furylacetone, furylacetone, 2-acetonylfuran, 2-propanone, 1-2-furanyl, furfuryl methyl ketone, 1-2-furyl-2-propanone, methyl furfuryl ketone, 1-2-furanyl-2-propanone, 2-furfuryl methyl ketone, 2-furyl-2-propanone
PubChem CID 228583
IUPAC Name 1-(furan-2-yl)propan-2-one
SMILES CC(=O)CC1=CC=CO1

Specifications

Boiling Point 179°C to 180°C
Flash Point 65°C (149°F)
Odor Stench
Quantity 5 g
Solubility Information Soluble in alcohol,triacetin.
Formula Weight 124.14
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Furylacetone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.