missing translation for 'onlineSavingsMsg'
Learn More

2-Heptanol, 99+%

Product Code. 10321374 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
50mL
This item is not returnable. View return policy

Product Code. 10321374

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 543-49-7 | C7H16O | 116.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 543-49-7
Molecular Formula C7H16O
Molecular Weight (g/mol) 116.20
MDL Number MFCD00004587
InChI Key CETWDUZRCINIHU-UHFFFAOYNA-N
Synonym 2-heptanol, s-heptyl alcohol, 2-heptyl alcohol, 2-hydroxyheptane, amyl methyl carbinol, heptanol-2, methyl amyl carbinol, 1-methylhexanol, heptyl alcohol, sec, 2-heptanol natural
PubChem CID 10976
IUPAC Name heptan-2-ol
SMILES CCCCCC(C)O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Color Colorless
Density 0.8175g/mL
Boiling Point 160°C to 162°C
Flash Point 59°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)4CH(OH)CH3
Refractive Index 1.4200 to 1.422
Quantity 1 L
Beilstein 01, 415
Merck Index 15, 4697
Specific Gravity 0.8175
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,ether,benzene,miscible with most common organic liquids
Formula Weight 116.2
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 2-Heptanol, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Heptanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H312-Harmful in contact with skin.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.