missing translation for 'onlineSavingsMsg'
Learn More

2-Heptanone, 98%

Product Code. 10051760 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
250 mL
1 L
5 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
100mL
250mL
5 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10051760 250 mL Glass bottle 250mL
10051750 100 mL Glass bottle 100mL
10307380 1 L Glass bottle 1 L
10237213 5 L Plastic drum 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10051760 Supplier Thermo Scientific Acros Supplier No. 154002500
  • ONLINE EXCLUSIVE PRICE
    69.85 €
    67.80 € / 250mL
    Save 2.05 €
    2% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 110-43-0 | C7H14O | 114.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-43-0
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.19
MDL Number MFCD00009513
InChI Key CATSNJVOTSVZJV-UHFFFAOYSA-N
Synonym 2-heptanone, methyl amyl ketone, butylacetone, methyl pentyl ketone, methyl n-amyl ketone, amyl methyl ketone, n-amyl methyl ketone, n-pentyl methyl ketone, heptanone, pentyl methyl ketone
PubChem CID 8051
ChEBI CHEBI:5672
IUPAC Name heptan-2-one
SMILES CCCCCC(C)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -35.0°C
Color Colorless
Density 0.8200g/mL
Boiling Point 149.0°C to 150.0°C
Flash Point 39°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3(CH2)4COCH3
Refractive Index 1.4075 to 1.4095
Quantity 250 mL
Beilstein 01, 699
Merck Index 15, 4698
Specific Gravity 0.82
Solubility Information Solubility in water: 4.3g/L (20°C). Other solubilities: soluble in alcohol and ether
Formula Weight 114.19
Percent Purity 98%
Grade Pure
Physical Form Liquid
Chemical Name or Material 2-Heptanone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Heptanone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.