missing translation for 'onlineSavingsMsg'
Learn More

2-Heptanone, 99%

Product Code. 11469512
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11469512 100 mL 100mL
11369609 250 mL 250mL
11479512 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11469512 Supplier Thermo Scientific Alfa Aesar Supplier No. A10200.AE
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 110-43-0 | C7H14O | 114.19 g/mol

2-Heptanone is used as a solvent for resins and lacquers. It is also used as a fragrance in cosmetics and a flavor for foods. It is involved as a reagent in the synthesis of rifamycin derivatives, which exhibit antibacterial activity. Further, it is employed in the preparation of thiazolidin-4-one derivatives as inhibitors of Toxoplasma gondii. In addition to this, it is considered as an alternative local anesthetic to lidocaine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Heptanone is used as a solvent for resins and lacquers. It is also used as a fragrance in cosmetics and a flavor for foods. It is involved as a reagent in the synthesis of rifamycin derivatives, which exhibit antibacterial activity. Further, it is employed in the preparation of thiazolidin-4-one derivatives as inhibitors of Toxoplasma gondii. In addition to this, it is considered as an alternative local anesthetic to lidocaine.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents, strong reducing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-43-0
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.19
MDL Number MFCD00009513
InChI Key CATSNJVOTSVZJV-UHFFFAOYSA-N
Synonym 2-heptanone, methyl amyl ketone, butylacetone, methyl pentyl ketone, methyl n-amyl ketone, amyl methyl ketone, n-amyl methyl ketone, n-pentyl methyl ketone, heptanone, pentyl methyl ketone
PubChem CID 8051
ChEBI CHEBI:5672
IUPAC Name heptan-2-one
SMILES CCCCCC(C)=O

Specifications

Melting Point -35°C
Density 0.82
Boiling Point 149°C to 150°C
Flash Point 47°C (116°F)
Odor Unpleasant
Refractive Index 1.409
Quantity 100 mL
UN Number UN1110
Beilstein 1699063
Merck Index 14,4663
Solubility Information Slightly miscible with water.
Formula Weight 114.19
Percent Purity 99%
Chemical Name or Material 2-Heptanone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Heptanone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 2-Heptanone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.