missing translation for 'onlineSavingsMsg'
Learn More

2-Heptyne, 98+%, Thermo Scientific™

Product Code. 11487094
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11487094 50 g 50g
11467094 2 g 2g
11477094 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11487094 Supplier Thermo Scientific Alfa Aesar Supplier No. B21528.18
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1119-65-9
Molecular Formula C7H12
Molecular Weight (g/mol) 96.173
MDL Number MFCD00039970
InChI Key AMSFEMSYKQQCHL-UHFFFAOYSA-N
Synonym 2-heptyne, 1-methyl-2-butylacetylene, butylmethylacetylene, n-butylmethylacetylene, acmc-1brzr, n-c4h9c.$.cch3, 2-heptyne,
PubChem CID 14245
IUPAC Name hept-2-yne
SMILES CCCCC#CC

Specifications

Density 0.753
Boiling Point 112°C to 113°C
Flash Point 11°C (52°F)
Odor Characteristic
Linear Formula CH3(CH2)3C≡CCH3
Refractive Index 1.422
Quantity 50 g
UN Number UN3295
Beilstein 1733563
Formula Weight 96.17
Percent Purity ≥98%
Chemical Name or Material 2-Heptyne
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.