missing translation for 'onlineSavingsMsg'
Learn More

2-Hexanone, Reagent grade, 98%, Honeywell™

Product Code. 15630970 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass Bottle
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 15630970

Brand: Honeywell Research Chemicals 10300410G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade, 98%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-78-6
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00009482
InChI Key QQZOPKMRPOGIEB-UHFFFAOYSA-N
Synonym 2-hexanone, hexanone, 2-oxohexane, propylacetone, n-butyl methyl ketone, butyl methyl ketone, methyl butyl ketone, methyl n-butyl ketone, hexanone-2, ketone, butyl methyl
PubChem CID 11583
IUPAC Name hexan-2-one
SMILES CCCCC(C)=O

Specifications

Melting Point -57°C
Color Colorless
Density 0.812g/mL at 25°C
Boiling Point 127°C
Flash Point 23°C (73.4°F)
Packaging Glass Bottle
Linear Formula CH3(CH2)3COCH3
Refractive Index n20/D 1.401(lit.)
Quantity 10 g
UN Number UN1224
Beilstein 1737676
Formula Weight 100.16g/mol
Percent Purity 98%
Grade Reagent
Physical Form Liquid
Chemical Name or Material 2-Hexanone
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.