missing translation for 'onlineSavingsMsg'
Learn More

2-Hexene, cis + trans, tech. 85%

Product Code. 15420328
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15420328

Brand: Thermo Scientific Alfa Aesar H53472.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 592-43-8 | C6H12 | 84.162 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 592-43-8
Molecular Formula C6H12
Molecular Weight (g/mol) 84.162
MDL Number MFCD00009473
InChI Key RYPKRALMXUUNKS-HWKANZROSA-N
Synonym cis-2-hexene, 2-hexene, z, z-2-hexene, z-hex-2-ene, 2-hexene, cis, unii-ui6o1h5j7j, 2-hexene-cis, 2z-hex-2-ene, ui6o1h5j7j, 2-hexene z
PubChem CID 639661
IUPAC Name (E)-hex-2-ene
SMILES CCCC=CC

Specifications

Density 0.678
Boiling Point 67°C to 69°C
Flash Point −20°C (−4°F)
Linear Formula CH3(CH2)2CH=CHCH3
Refractive Index 1.395
Quantity 5 g
UN Number UN3295
Beilstein 969179
Formula Weight 84.16
Percent Purity 85%
Grade Technical
Chemical Name or Material 2-Hexene, Cis + Trans
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.