missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-2-propanesulfonic acid monosodium salt, 97%

Product Code. 11343318
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11343318

Brand: Thermo Scientific Alfa Aesar 036256.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 540-92-1 | C3H8NaO4S+ | 163.143 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540-92-1
Molecular Formula C3H8NaO4S+
Molecular Weight (g/mol) 163.143
MDL Number MFCD00031395
InChI Key YNJORDSKPXMABC-UHFFFAOYSA-N
Synonym sodium acetone bisulfite, acetone-sodium bisulfite adduct, sodium bisulfite-acetone adduct, acetone-monosodium sulfite adduct, sodium 2-hydroxy-2-propane-sulfonate, acetone-sodium bisulfite compound 1:1, acetone, compound with sodium bisulfite 1:1, 2-propanone, compd. with monosodium sulfite 1:1, 2-propanesulfonic acid, monosodium salt, sodium 2-hydroxypropane-2-sulfonic acid
PubChem CID 54602011
IUPAC Name sodium;2-hydroxypropane-2-sulfonic acid
SMILES CC(C)(O)S(=O)(=O)O.[Na+]

Specifications

Quantity 100 g
Formula Weight 162.14
Percent Purity 97%
Physical Form Crystalline
Chemical Name or Material 2-Hydroxy-2-propanesulfonic acid monosodium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.