missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-2-(trifluoromethyl)propionic acid, 94%

Product Code. 11348396
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11348396 5 g 5g
11338396 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11348396 Supplier Thermo Scientific Alfa Aesar Supplier No. L17399.06
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 374-35-6 | C4H5F3O3 | 158.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 374-35-6
Molecular Formula C4H5F3O3
Molecular Weight (g/mol) 158.08
MDL Number MFCD00190646,MFCD03095400,MFCD07375449
InChI Key CTGJACFEVDCYMC-UHFFFAOYNA-N
Synonym 2-trifluoromethyl-2-hydroxypropionic acid, 3,3,3-trifluoro-2-hydroxy-2-methylpropionic acid, 2-hydroxy-2-trifluoromethyl propionic acid, 3,3,3-trifluoro-2-hydroxy-2-methyl-propionic acid, propanoic acid, 3,3,3-trifluoro-2-hydroxy-2-methyl, methyl3,3,3-trifluoro-dl-lactate, acmc-1ai3k, acmc-209gc9, 2s-2-hydroxy-2-methyl-3,3,3-trifluoropropanoic acid, alpha-trifluoromethyl lactic acid
PubChem CID 2775125
IUPAC Name 3,3,3-trifluoro-2-hydroxy-2-methylpropanoic acid
SMILES CC(O)(C(O)=O)C(F)(F)F

Specifications

Melting Point ∼85°C
Boiling Point 189°C
Quantity 5 g
UN Number UN3261
Beilstein 1723691
Formula Weight 158.08
Percent Purity 94%
Chemical Name or Material 2-Hydroxy-2-(trifluoromethyl)propionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.