missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-3-(4-(4-hydroxy-3,5-diiodophenoxy)-3,5-dinitrophenyl)propanoic acid, TRC

Product Code. 30401568
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30401568 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30401568 Supplier Toronto Research Chemicals Supplier No. TRCH942968250MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Hydroxy-3-(4-(4-hydroxy-3,5-diiodophenoxy)-3,5-dinitrophenyl)propanoic acid
CAS 2514752-23-7
Molecular Formula C15H10I2N2O9
Quantity 250 mg
InChI Formula InChI=1S/C15H10I2N2O9/c16-8-4-7(5-9(17)13(8)21)28-14-10(18(24)25)1-6(2-11(14)19(26)27)3-12(20)15(22)23/h1-2,4-5,12,20-21H,3H2,(H,22,23)
SMILES O=C(O)C(O)CC1=CC([N+]([O-])=O)=C(OC2=CC(I)=C(O)C(I)=C2)C([N+]([O-])=O)=C1
IUPAC Name 2-hydroxy-3-(4-(4-hydroxy-3,5-diiodophenoxy)-3,5-dinitrophenyl)propanoic acid
Molecular Weight (g/mol) 616.06
Formula Weight 615.85
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.