missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-3-methylbutyric acid, 97%

Product Code. 15427378
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15427378 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15427378 Supplier Thermo Scientific Alfa Aesar Supplier No. H60872.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4026-18-0 | C5H10O3 | 118.132 g/mol

2-Hydroxy-3-methylbutyric acid is employed as internal standard for analysis of ethylene glycol (EG) and γ-hydroxybutyrate (GHB) in whole blood.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxy-3-methylbutyric acid is employed as internal standard for analysis of ethylene glycol (EG) and γ-hydroxybutyrate (GHB) in whole blood.

Solubility
Soluble in dimethyl sulfoxide and methanol.

TRUSTED_SUSTAINABILITY

Specifications

CAS 4026-18-0
Melting Point 82°C to 84°C
Molecular Formula C5H10O3
Linear Formula (CH3)2CHCH(OH)COOH
MDL Number MFCD00004242
Quantity 1 g
Beilstein 1721138
Synonym 2-hydroxy-3-methylbutyric acid, 2-hydroxyisovaleric acid, alpha-hydroxyisovaleric acid, 2-oxyisovaleric acid, 3-methyl-2-hydroxybutyric acid, butanoic acid, 2-hydroxy-3-methyl, 2-hydroxyisopentanoic acid, dl-2-hydroxyisovaleric acid, dl-2-hydroxy-3-methylbutanoic acid, 2-hydroxyisopentanoate
Solubility Information Soluble in dimethyl sulfoxide and methanol.
InChI Key NGEWQZIDQIYUNV-UHFFFAOYSA-N
SMILES CC(C)C(C(=O)O)O
IUPAC Name 2-hydroxy-3-methylbutanoic acid
Molecular Weight (g/mol) 118.132
PubChem CID 99823
ChEBI CHEBI:60645
Formula Weight 118.13
Percent Purity 98%
Chemical Name or Material 2-Hydroxy-3-methylbutyric acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Hydroxy-3-methylbutyric acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 2-Hydroxy-3-methylbutyric acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.