missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-4-iodobenzaldehyde, TRC

Product Code. 30391799
Change view
Click to view available options
Quantity:
500 mg
2.5 g
5 g
Unit Size:
2.50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391799 2.5 g 2.50g
30392711 5 g -
30397498 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30391799 Supplier Toronto Research Chemicals Supplier No. TRCH9563482.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38170-02-4
Molecular Formula C7H5IO2
Molecular Weight (g/mol) 248.02
Synonym 4-Iodo-2-hydroxybenzaldehyde, 4-Iodosalicylaldehyde, 4-Iodo-salicylaldehyde
IUPAC Name 2-hydroxy-4-iodobenzaldehyde
SMILES Oc1cc(I)ccc1C=O

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C7H5IO2/c8-6-2-1-5(4-9)7(10)3-6/h1-4,10H
Formula Weight 247.9334
Chemical Name or Material 2-Hydroxy-4-iodobenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.