missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-4-methylbenzaldehyde, 97+%

Product Code. 15466797
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15466797 25 g 25g
15456797 5 g 5g
15476797 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15466797 Supplier Thermo Scientific Alfa Aesar Supplier No. H33909.14
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 698-27-1 | C8H8O2 | 136.15 g/mol

The intermediate for synthesis of liquid crystal. Also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The intermediate for synthesis of liquid crystal. Also used as pharmaceutical intermediate.

Solubility
Soluble in oil, slightly soluble in water.

Notes
Air sensitive. Store away from air, oxidizing agents. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 698-27-1
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00799550
InChI Key JODRRPJMQDFCBJ-UHFFFAOYSA-N
Synonym 4-methylsalicylaldehyde, 2,4-cresotaldehyde, benzaldehyde, 2-hydroxy-4-methyl, 4-methyl-2-hydroxybenzaldehyde, 2-hydroxy-4-methyl-benzaldehyde, 2-formyl-5-methylphenol, 4-methylsalicyclic aldehyde, unii-9n0s26g61j, 4-methyl salicylaldehyde, homosalicylaldehyde, m
PubChem CID 61200
IUPAC Name 2-hydroxy-4-methylbenzaldehyde
SMILES CC1=CC=C(C=O)C(O)=C1

Specifications

Melting Point 58°C to 62°C
Boiling Point 222°C to 223°C
Quantity 25 g
Sensitivity Air Sensitive
Solubility Information Soluble in oil,slightly soluble in water.
Formula Weight 136.15
Percent Purity ≥97%
Chemical Name or Material 2-Hydroxy-4-methylbenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.