missing translation for 'onlineSavingsMsg'
Learn More

2-(((Hydroxy-4-methyloctyl)oxy)carbonyl)cyclohexanecarboxylic Acid (Mixture of Diastereomers), TRC

Product Code. 17321820
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17321820 10 mg 10mg
17949264 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17321820 Supplier Toronto Research Chemicals Supplier No. TRCH94971010MG
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1637562-52-7
Molecular Formula C17H30O5
Molecular Weight (g/mol) 314.42
Synonym 1,2-Cyclohexanedicarboxylic Acid, Mono-4-methyl-7-hydroxyoctyl Ester, 1-(7-Hydroxy-4-methyloctyl) 1,2-cyclohexanedicarboxylate, Cyclohexane-1,2-dicarboxylate mono-(7-hydroxy-4-methyloctyl) ester, 1,2-Cyclohexanedicarboxylic acid, 1-(7-hydroxy-4-methyloctyl) ester
IUPAC Name 2-(7-hydroxy-4-methyl-octoxy)carbonylcyclohexanecarboxylic acid
SMILES CC(O)CCC(C)CCCOC(=O)C1CCCCC1C(=O)O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C17H30O5/c1-12(9-10-13(2)18)6-5-11-22-17(21)15-8-4-3-7-14(15)16(19)20/h12-15,18H,3-11H2,1-2H3,(H,19,20)
Formula Weight 314.2093
Percent Purity >95%
Chemical Name or Material Cyclohexane-1,2-dicarboxylic acid, mono (7-hydroxy- 4-methyloctyl)ester
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.