missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-4-methylvaleric acid, 98%

Product Code. 15456928
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15456928 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15456928 Supplier Thermo Scientific Alfa Aesar Supplier No. H60459.03
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 498-36-2 | C6H12O3 | 132.159 g/mol

2-Hydroxy-4-methylvaleric acid is used as nutrients, stabilizers, surfactants and emulsifiers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxy-4-methylvaleric acid is used as nutrients, stabilizers, surfactants and emulsifiers.

Solubility
Soluble in water. (82 g/L) at 25°C.

Notes
Store in a cool, dry conditions in a well seal
TRUSTED_SUSTAINABILITY

Specifications

CAS 498-36-2
Melting Point 78°C to 80°C
Molecular Formula C6H12O3
Linear Formula (CH3)2CHCH2CH(OH)CO2H
MDL Number MFCD00004246
Quantity 1 g
Synonym 2-hydroxy-4-methylvaleric acid, 2-hydroxyisocaproic acid, leucic acid, dl-leucic acid, alpha-hydroxyisocaproic acid, leucinic acid, 2-hydroxy-4-methyl-pentanoic acid, pentanoic acid, 2-hydroxy-4-methyl, dl-2-hydroxyisocaproic acid, leucate
Solubility Information Soluble in water. (82g/L) at 25°C.
InChI Key LVRFTAZAXQPQHI-UHFFFAOYSA-N
SMILES CC(C)CC(C(=O)O)O
IUPAC Name 2-hydroxy-4-methylpentanoic acid
Molecular Weight (g/mol) 132.159
PubChem CID 92779
ChEBI CHEBI:59783
Formula Weight 132.16
Percent Purity 98%
Chemical Name or Material 2-Hydroxy-4-methylvaleric acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.