missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-5-[2-(4-hydroxyphenyl)propan-2-yl]benzoic Acid, TRC

Product Code. 30396402
Change view
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
100mg
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30396402 100 mg 100mg
30399768 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30396402 Supplier Toronto Research Chemicals Supplier No. TRCH953975100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Hydroxy-5-[2-(4-hydroxyphenyl)propan-2-yl]benzoic Acid
CAS 101949-49-9
Molecular Formula C16H16O4
Quantity 100 mg
InChI Formula InChI=1S/C16H16O4/c1-16(2,10-3-6-12(17)7-4-10)11-5-8-14(18)13(9-11)15(19)20/h3-9,17-18H,1-2H3,(H,19,20)
SMILES CC(C)(c1ccc(O)cc1)c2ccc(O)c(c2)C(=O)O
IUPAC Name 2-hydroxy-5-[2-(4-hydroxyphenyl)propan-2-yl]benzoic acid
Molecular Weight (g/mol) 272.3
Formula Weight 272.1049
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.