missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-6-methylisonicotinic acid, 97%, Thermo Scientific™

Product Code. 10482304
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10482304 10 g 10g
10262642 1 g 1g
10479454 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10482304 Supplier Thermo Scientific Maybridge Supplier No. MO00027EA
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 86454-13-9
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.14
MDL Number MFCD02682019
InChI Key NQXYVTIQDNOHFM-UHFFFAOYSA-N
Synonym 2-hydroxy-6-methylisonicotinic acid, 2-hydroxy-6-methylpyridine-4-carboxylic acid, 6-methyl-2-oxo-1,2-dihydropyridine-4-carboxylic acid, 2-hydroxy-6-methyl-isonicotinicacid, 6-hydroxy-2-methylpyridine-4-carboxylic acid, 1,2-dihydro-6-methyl-2-oxopyridine-4-carboxylic acid, 4-pyridinecarboxylic acid, 1,2-dihydro-6-methyl-2-oxo, 4-pyridinecarboxylicacid, 1,2-dihydro-6-methyl-2-oxo, maybridge3_004231, acmc-209q9q
PubChem CID 276660
IUPAC Name 2-methyl-6-oxo-1H-pyridine-4-carboxylic acid
SMILES CC1=CC(=CC(=O)N1)C(O)=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 10 g
Formula Weight 153.14
Percent Purity 97%
Chemical Name or Material 2-Hydroxy-6-methylisonicotinic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.