missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-6-methylpyridine-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 11312839
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11312839 1 g 1g
11322839 5 g 5g
11332839 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11312839 Supplier Thermo Scientific Alfa Aesar Supplier No. H27716.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86454-13-9
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.14
MDL Number MFCD02682019
InChI Key NQXYVTIQDNOHFM-UHFFFAOYSA-N
Synonym 2-hydroxy-6-methylisonicotinic acid, 2-hydroxy-6-methylpyridine-4-carboxylic acid, 6-methyl-2-oxo-1,2-dihydropyridine-4-carboxylic acid, 2-hydroxy-6-methyl-isonicotinicacid, 6-hydroxy-2-methylpyridine-4-carboxylic acid, 1,2-dihydro-6-methyl-2-oxopyridine-4-carboxylic acid, 4-pyridinecarboxylic acid, 1,2-dihydro-6-methyl-2-oxo, 4-pyridinecarboxylicacid, 1,2-dihydro-6-methyl-2-oxo, maybridge3_004231, acmc-209q9q
PubChem CID 276660
IUPAC Name 2-methyl-6-oxo-1H-pyridine-4-carboxylic acid
SMILES CC1=CC(=CC(=O)N1)C(O)=O

Specifications

Melting Point >300°C (decomposition)
Quantity 1 g
Formula Weight 153.15
Percent Purity 97%
Chemical Name or Material 2-Hydroxy-6-methylpyridine-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.