missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy Desipramine, TRC

Product Code. 30400738
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30400738 1 mg 1mg
30406913 10 mg -
30408289 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30400738 Supplier Toronto Research Chemicals Supplier No. TRCH9365001MG
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1977-15-7
Molecular Formula C18 H22 N2 O
Molecular Weight (g/mol) 282.38
Synonym 5H-Dibenz[b,f]azepin-2-ol, 10,11-dihydro-5-[3-(methylamino)propyl]-, 10,11-Dihydro-5-[3-(methylamino)propyl]-5H-dibenz[b,f]azepin-2-ol, 2-Hydroxydemethylimipramine, 2-Hydroxydesipramine, 2-Hydroxydesmethylimipramine, GP 36329, Hydroxydesipramine
IUPAC Name 11-[3-(methylamino)propyl]-5,6-dihydrobenzo[b][1]benzazepin-3-ol
SMILES CNCCCN1c2ccccc2CCc3cc(O)ccc13

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C18H22N2O/c1-19-11-4-12-20-17-6-3-2-5-14(17)7-8-15-13-16(21)9-10-18(15)20/h2-3,5-6,9-10,13,19,21H,4,7-8,11-12H2,1H3
Formula Weight 282.1732
Chemical Name or Material 2-Hydroxy Desipramine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.