missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-gamma-butyrolactone, 99%

Product Code. 10581121 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10581121 25 g Glass bottle 25g
10333722 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10581121 Supplier Thermo Scientific Acros Supplier No. 275070250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 19444-84-9 | C4H6O3 | 102.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19444-84-9
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.09
MDL Number MFCD00134268,MFCD00211245
InChI Key FWIBCWKHNZBDLS-UHFFFAOYNA-N
Synonym alpha-hydroxy-gamma-butyrolactone, 3-hydroxydihydrofuran-2 3h-one, 2-hydroxy-gamma-butyrolactone, 3-hydroxytetrahydrofuran-2-one, 2 3h-furanone, dihydrohydroxy, 3s-3-hydroxydihydrofuran-2 3h-one, 3-hydroxydihydro-2 3h-furanone, hydroxybutyrolactone, r-dihydro-3-hydroxyfuran-2 3h-one, 2 3h-furanone, dihydro-3-hydroxy
PubChem CID 545831
IUPAC Name 3-hydroxyoxolan-2-one
SMILES OC1CCOC1=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.3130g/mL
Boiling Point 85°C to 90°C (0.8 mbar)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4760 to 1.481
Quantity 25 g
Beilstein 18, V,1, 3
Specific Gravity 1.313
Formula Weight 102.09
Percent Purity 99%
Physical Form Viscous Liquid
Chemical Name or Material 2-Hydroxy-γ-butyrolactone, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Hydroxy-y-butyrolactone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.