missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy Imipramine, TRC

Product Code. 30137784
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137784 10 mg 10mg
17910455 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137784 Supplier Toronto Research Chemicals Supplier No. TRCH94340010MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 303-70-8
Molecular Formula C19 H24 N2 O
Molecular Weight (g/mol) 296.41
IUPAC Name 11-[3-(dimethylamino)propyl]-5,6-dihydrobenzo[b][1]benzazepin-3-ol
SMILES CN(C)CCCN1c2ccccc2CCc3cc(O)ccc13

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C19H24N2O/c1-20(2)12-5-13-21-18-7-4-3-6-15(18)8-9-16-14-17(22)10-11-19(16)21/h3-4,6-7,10-11,14,22H,5,8-9,12-13H2,1-2H3
Formula Weight 296.1889
Chemical Name or Material 2-Hydroxy Imipramine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.