missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxybenzylamine, TRC

Product Code. 30403952
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
30403952 1 g 1g
1 options
This item is not returnable. View return policy

Product Code. 30403952

Brand: Toronto Research Chemicals TRCH8097251G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(Aminomethyl)phenol
CAS 932-30-9
Molecular Formula C7 H9 N O
Quantity 1 g
Synonym Phenol, 2-(aminomethyl)-, o-Cresol, α-amino- (6CI,7CI,8CI), 2-(Aminomethyl)phenol, 2-Hydroxybenzenemethanamine, 2-Hydroxybenzylamine, NSC 127870, Salicylamine, o-Hydroxybenzylamine
InChI Formula InChI=1S/C7H9NO/c8-5-6-3-1-2-4-7(6)9/h1-4,9H,5,8H2
SMILES NCc1ccccc1O
IUPAC Name 2-(aminomethyl)phenol
Molecular Weight (g/mol) 123.153
Formula Weight 123.068
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.