missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxycaprylic Acid, TRC

Product Code. 30395147
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30395147 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30395147 Supplier Toronto Research Chemicals Supplier No. TRCH999988500MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Hydroxycaprylic Acid
CAS 617-73-2
Molecular Formula C8H16O3
Quantity 500 mg
Synonym 2-Hydroxycaprylic Acid, Octanoic acid, 2-hydroxy-, 2-Hydroxyoctanoic acid, (±)-2-Hydroxyoctanoic acid, (±)-α-Hydroxycaprylic acid, 2-Hydroxycaprylic acid, DL-2-Hydroxycaprylic acid, DL-2-Hydroxyoctanoic acid, NSC 189703, α-Hydroxy-n-caprylic acid, α-Hydroxycaprylic acid, α-Hydroxyoctanoic acid, 2-Hydroxycaprylic Acid, Octanoic acid, 2-hydroxy-, 2-Hydroxyoctanoic acid, (±)-2-Hydroxyoctanoic acid, (±)-α-Hydroxycaprylic acid, 2-Hydroxycaprylic acid, DL-2-Hydroxycaprylic acid, DL-2-Hydroxyoctanoic acid, NSC 189703, α-Hydroxy-n-caprylic acid, α-Hydroxycaprylic acid, α-Hydroxyoctanoic acid
InChI Formula InChI=1S/C8H16O3/c1-2-3-4-5-6-7(9)8(10)11/h7,9H,2-6H2,1H3,(H,10,11)
SMILES CCCCCCC(O)C(=O)O
IUPAC Name 2-hydroxyoctanoic acid
Molecular Weight (g/mol) 160.21
Formula Weight 160.1099
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.