missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxycarbazole, 97%

Product Code. 11372929
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11372929 5 g 5g
11382929 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11372929 Supplier Thermo Scientific Alfa Aesar Supplier No. H27765.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 86-79-3 | C12H9NO | 183.21 g/mol

Carbazol has been found possible to distinguish between heparin, heparin derivatives, and other polyuronides of connective tissue. A new modification of the carbazole analysis is useful for determination of d-mannuronic acid in heteropolysaccharides. 2-Hydroxycarbazole was used in the synthesis of isochromene fused carbazol, (4aS,13bR)-2,5,5-trimethyl-3,4,4a,5,8,13b-hexahydroisochromeno[3,4-b]carbazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Carbazol has been found possible to distinguish between heparin, heparin derivatives, and other polyuronides of connective tissue. A new modification of the carbazole analysis is useful for determination of d-mannuronic acid in heteropolysaccharides. 2-Hydroxycarbazole was used in the synthesis of isochromene fused carbazol, (4aS,13bR)-2,5,5-trimethyl-3,4,4a,5,8,13b-hexahydroisochromeno[3,4-b]carbazole.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-79-3
Molecular Formula C12H9NO
Molecular Weight (g/mol) 183.21
MDL Number MFCD00004962
InChI Key GWPGDZPXOZATKL-UHFFFAOYSA-N
Synonym 2-hydroxycarbazole, carbazol-2-ol, ccris 5301, 2-hydroxycarbazol, 7-hydroxycarbazole, 2-hydroxy carbazole, pubchem9945, pubchem9949, 2-hydroxy-9h-carbazol, 2-hydroxy-9h-carbazole
PubChem CID 93551
IUPAC Name 9H-carbazol-2-ol
SMILES OC1=CC2=C(C=C1)C1=C(N2)C=CC=C1

Specifications

Melting Point 262°C to 273°C
Odor Odorless
Quantity 5 g
Formula Weight 183.21
Percent Purity 97%
Chemical Name or Material 2-Hydroxycarbazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.