missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxyisobutyric acid, 98%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Plastic bottle
Unit Size:
100g
1kg
25g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS Min % | 97.5 |
| CAS Max % | 100.0 |
| Melting Point | 77°C to 81°C |
| Boiling Point | 84°C (1.5 mmHg) |
| Assay Percent Range | 98% |
| Packaging | Plastic bottle |
| Linear Formula | (CH3)2C(OH)CO2H |
| Quantity | 25 g |
| Beilstein | 03, 313 |
| Solubility Information | Solubility in water: 1900g/L (22°C). Other solubilities: soluble in methanol,diethyl ether and hot benzene,sparingly soluble in cold benzene |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction