missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxymethyl-15-crown-5, 98%

Product Code. 11495210
Change view
Click to view available options
Quantity:
0.25 g
1 g
5 g
Unit Size:
0.25g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495210 5 g 5g
11475210 0.25 g 0.25g
11485210 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11495210 Supplier Thermo Scientific Alfa Aesar Supplier No. 044500.06
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 75507-25-4 | C11H22O6 | 250.291 g/mol

2-Hydroxymethyl-15-crown-5 is used for chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75507-25-4
Molecular Formula C11H22O6
Molecular Weight (g/mol) 250.291
MDL Number MFCD00192215
InChI Key YHIQMMGCRYKJLB-UHFFFAOYSA-N
PubChem CID 2733744
IUPAC Name 1,4,7,10,13-pentaoxacyclopentadec-2-ylmethanol
SMILES C1COCCOCC(OCCOCCO1)CO

Specifications

Density 1.175 g/mL
Boiling Point 135°C (1.5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.479
Quantity 5 g
Beilstein 3589706
Synonym 1,4,7,10,13-pentaoxacyclopentadecane-2-methanol, 2-hydroxymethyl-15-crown-5, 2-hydroxymethyl-15-crown 5-ether, 1,4,7,10,13-pentaoxacyclopentadecan-2-ylmethanol, hydroxy-15-crown-5, 2-hydroxymethyl-15-crown5-ether, 2-hydroxymethyl-15-crown, 2,5,8,11,14-pentaoxacyclopentadecyl methan-1-ol, 1,4,7,10,13-pentaoxacyclopentadec-2-yl-methanol, 1,4,7,10,13-pentaoxacyclopentadecan-2-yl methanol
Solubility Information Soluble in water(58g/L) (25°C).
Formula Weight 250.29
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Hydroxymethyl-15-crown-5
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.