missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyphenone, TRC

Product Code. 30402000
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30402000 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30402000 Supplier Toronto Research Chemicals Supplier No. TRCH8293205G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Hydroxyphenone
CAS 117-99-7
Molecular Formula C13H10O2
Quantity 5 g
Synonym 2-Hydroxybenzophenone, (2-Hydroxyphenyl)phenylmethanone, 2-Benzoylphenol2-Hydroxybenzophenone, 2'-Hydroxybenzophenone, NSC 623, Phenyl 2-Hydroxyphenyl Ketone, UVA 635LP, o-Benzoylphenol, o-Hydroxybenzophenoneo-Hydroxyphenyl Phenyl Ketone
InChI Formula InChI=1S/C13H10O2/c14-12-9-5-4-8-11(12)13(15)10-6-2-1-3-7-10/h1-9,14H
SMILES C(C1=CC=CC=C1O)(C1=CC=CC=C1)=O
IUPAC Name (2-hydroxyphenyl)-phenylmethanone
Molecular Weight (g/mol) 198.22
Formula Weight 198.0681
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.