missing translation for 'onlineSavingsMsg'
Learn More

(2-Hydroxypropyl)-beta-cyclodextrin, MW ca 1250-1480

Product Code. 15414047
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15414047 5 g 5g
15437795 25 g 25g
15447795 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15414047 Supplier Thermo Scientific Alfa Aesar Supplier No. H31133.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 128446-35-5 | C44H75O36 | 1180.05 g/mol

2-Hydroxypropyl)-β-cyclodextrin is used to enhance solubility of hydrophobic compounds by forming complex with them. This favors its usage in pharmaceutics, in the manufacturing of drugs. Beta- cyclodextrin and its methyl derivative is used to remove cholesterol from cultured cells. Many common chiral stationary phases used in high-performance liquid chromatography columns consists of beta-cyclodextrin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxypropyl)-β-cyclodextrin is used to enhance solubility of hydrophobic compounds by forming complex with them. This favors its usage in pharmaceutics, in the manufacturing of drugs. Beta- cyclodextrin and its methyl derivative is used to remove cholesterol from cultured cells. Many common chiral stationary phases used in high-performance liquid chromatography columns consists of beta-cyclodextrin.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents. Keep the container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128446-35-5
Molecular Formula C44H75O36
Molecular Weight (g/mol) 1180.05
MDL Number MFCD00069372

Specifications

Melting Point 305°C
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight ≈1250 to 1480 d
Chemical Name or Material (2-Hydroxypropyl)-beta-cyclodextrin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.