missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxypyridine-4-carboxylic acid, 98%

Product Code. 15401349
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. Quantity unitSize
15401349 25 g 25g
15491339 5 g 5g
15411349 100 g 100g
3 options
This item is not returnable. View return policy

Product Code. 15401349

Brand: Thermo Scientific Alfa Aesar H64241.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 22282-72-0 | C6H5NO3 | 139.11 g/mol

Used in the synthesis of selective inhibitors of neuropeptide Y Y5 inhibiting food intake. Also used in the optimization of p-subsite residues of HIV protease inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the synthesis of selective inhibitors of neuropeptide Y Y5 inhibiting food intake. Also used in the optimization of p-subsite residues of HIV protease inhibitors.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22282-72-0
Molecular Formula C6H5NO3
Molecular Weight (g/mol) 139.11
MDL Number MFCD01318566
InChI Key BXHCJLRTXPHUGH-UHFFFAOYSA-N
Synonym 2-hydroxyisonicotinic acid, 2-hydroxy-4-pyridinecarboxylic acid, 2-oxo-1,2-dihydropyridine-4-carboxylic acid, 2-hydroxypyridine-4-carboxylic acid, 2-hydroxy-isonicotinic acid, 2-hydroxyisonicotinicacid, 4-pyridinecarboxylic acid, 1,2-dihydro-2-oxo, pubchem16780, acmc-209yjf, 2-hydroxy isonicotinic acid
PubChem CID 280756
SMILES OC(=O)C1=CC(=O)NC=C1

Specifications

Quantity 25 g
Beilstein 115859
IUPAC Name 2-oxo-1,2-dihydropyridine-4-carboxylic acid
Formula Weight 139.11
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Hydroxypyridine-4-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.