missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxypyridine, TRC

Product Code. 30394234
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30394234 10 g
30402154 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30394234 Supplier Toronto Research Chemicals Supplier No. TRCH95317010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72762-00-6
Molecular Formula C5 H5 N O
Molecular Weight (g/mol) 95.1
Synonym Pyridin-2-ol, Levetiracetam Imp. C (EP), 2(1H)-Pyridone (6CI,8CI), 2-Pyridol (3CI), 1,2-Dihydro-2-oxopyridine, 1H-2-Pyridone, 1H-Pyridin-2-one, 2-Hydroxypyridine, 2-Oxopyridine, 2-Pyridinol, 2-Pyridinone, 2-Pyridone, NSC 172522, Pyridin-2-one, α-Hydroxypyridine, α-Pyridone
IUPAC Name pyridin-2-ol
SMILES Oc1ccccn1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C5H5NO/c7-5-3-1-2-4-6-5/h1-4H,(H,6,7)
Formula Weight 95.0371
Chemical Name or Material Pyridin-2-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.