missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyquinoline-3-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10672475
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10672475 1 g Amber Glass Bottle 1g
10386424 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10672475 Supplier Thermo Scientific Maybridge Supplier No. MO08356DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 2003-79-4
Molecular Formula C10H7NO3
Molecular Weight (g/mol) 189.17
MDL Number MFCD01550054
InChI Key XOQQVKDBGLYPGH-UHFFFAOYSA-N
Synonym 2-hydroxyquinoline-3-carboxylic acid, 2-oxo-1,2-dihydroquinoline-3-carboxylic acid, 2-oxo-1,2-dihydro-quinoline-3-carboxylic acid, 2-oxoquinoline-3-carboxylic acid, 2-hydroxy-3-quinolinecarboxylic acid, 3-quinolinecarboxylic acid, 1,2-dihydro-2-oxo, 2-oxo-1,2-dihydro-3-quinolinecarboxylic acid, 1,2-dihydro-2-oxoquinoline-3-carboxylic acid, 3-carboxycarbostyril, oxychinolincarbonsaure
PubChem CID 332488
IUPAC Name 2-oxo-1H-quinoline-3-carboxylic acid
SMILES C1=CC=C2C(=C1)C=C(C(=O)N2)C(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 189.17
Percent Purity 97%
Chemical Name or Material 2-Hydroxyquinoline-3-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.