missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ 2-Hydroxysaclofen

Product Code. 18075442
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18075442 50 mg 50mg
18035922 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18075442 Supplier R&D Systems™ Supplier No. 0245/50
  • ONLINE EXCLUSIVE PRICE
    1268.65 €
    1142.40 € / 50mg
    Save 126.25 €
    9% Off
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective GABAB antagonist; more potent than Saclofen (Cat. No. 0246)

Selective antagonist at GABAB receptors.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117354-64-0
Molecular Formula C9H12ClNO4S

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 265.71
Purity >98%
Molecular Weight (g/mol) 265.708
Chemical Name or Material 2-Hydroxysaclofen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.