missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxysaclofen, Thermo Scientific Chemicals

Product Code. 15415789
Change view
Click to view available options
Quantity:
10 mg, 10mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15415789 10 mg, 10mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15415789 Supplier Thermo Scientific Alfa Aesar Supplier No. J63588.MA
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selective antagonist at GABA-B receptors

Selective antagonist at GABA-B receptors

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117354-64-0
Molecular Formula C9H12ClNO4S
Molecular Weight (g/mol) 265.708
MDL Number MFCD00069282
InChI Key WBSMZVIMANOCNX-UHFFFAOYSA-N
Synonym 2-hydroxysaclofen, 2-oh-saclofen, 2-hydroxy-saclofen, 3-amino-2-4-chlorophenyl-2-hydroxypropane-1-sulfonic acid, 2-hydroxy-s---saclofen, 3-amino-2-4-chlorophenyl-2-hydroxypropylsulfonic acid, 3-amino-2-4-chlorophenyl-2-hydroxypropanesulfonic acid, beta-aminomethyl-4-chloro-beta-hydroxybenzeneethanesulfonic acid, benzeneethanesulfonic acid, beta-aminomethyl-4-chloro-beta-hydroxy, 2-hydroxysaclofen, solid
PubChem CID 1564
IUPAC Name 3-amino-2-(4-chlorophenyl)-2-hydroxypropane-1-sulfonic acid
SMILES C1=CC(=CC=C1C(CN)(CS(=O)(=O)O)O)Cl

Specifications

Melting Point 268°C to 271°C
Color White
Quantity 10 mg, 10mg
Physical Form Solid
UN Number UN1759
Solubility Information Soluble in water at 1mg/ml; Also soluble in DMSO,0;1M HCl and methanol
Formula Weight 265.71
Chemical Name or Material 2-Hydroxysaclofen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.