missing translation for 'onlineSavingsMsg'
Learn More

2-Imidazolidinethione, 98%

Product Code. 10172870 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10172870 250 g Glass bottle 250g
10745381 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10172870 Supplier Thermo Scientific Acros Supplier No. 122032500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 96-45-7 | C3H6N2S | 102.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 96-45-7
Molecular Formula C3H6N2S
Molecular Weight (g/mol) 102.16
MDL Number MFCD00005276
InChI Key PDQAZBWRQCGBEV-UHFFFAOYSA-N
Synonym 2-imidazolidinethione, ethylenethiourea, ethylene thiourea, 2-mercaptoimidazoline, imidazolidinethione, n,n'-ethylenethiourea, mercazin i, warecure c, rhenogran etu, mercaptoimidazoline
PubChem CID 2723650
ChEBI CHEBI:34750
IUPAC Name imidazolidine-2-thione
SMILES S=C1NCCN1

Specifica

CAS Min % 97.5
CAS Max % 100.0
Melting Point 197.0°C to 201.0°C
Color White
Flash Point 252°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 250 g
Beilstein 24, 4
Merck Index 15, 3858
Solubility Information Solubility in water: 19g/L (20°C). Other solubilities: 440g/L water (90°C), moderately soluble in methanol, ethanol, ethylene, glycol and pyridine, insoluble in acetone, ether, chloroform, benzene, and ligroin
Formula Weight 102.15
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Imidazolidinethione, 98%
Vedi altri risultati Mostra meno risultati
compliance-icons compliance-icons
Identificativo prodotto
  • 2-Imidazolidinethione
Avvertenza
  • Danger
Categoria di pericolo
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1B
Indicazioni di pericolo
  • H360D-May damage the unborn child.
  • H302-Harmful if swallowed.
Consigli di prudenza
  • P201-Obtain special instructions before use.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.