missing translation for 'onlineSavingsMsg'
Learn More

2-Imidazolidinethione, 98%

Product Code. 10745381 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10745381 1 kg Plastic bottle 1kg
10172870 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10745381 Supplier Thermo Scientific Acros Supplier No. 122030010
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 96-45-7 | C3H6N2S | 102.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-45-7
Molecular Formula C3H6N2S
Molecular Weight (g/mol) 102.16
MDL Number MFCD00005276
InChI Key PDQAZBWRQCGBEV-UHFFFAOYSA-N
Synonym 2-imidazolidinethione, ethylenethiourea, ethylene thiourea, 2-mercaptoimidazoline, imidazolidinethione, n,n'-ethylenethiourea, mercazin i, warecure c, rhenogran etu, mercaptoimidazoline
PubChem CID 2723650
ChEBI CHEBI:34750
IUPAC Name imidazolidine-2-thione
SMILES S=C1NCCN1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 197.0°C to 201.0°C
Color White
Flash Point 252°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Plastic bottle
Quantity 1 kg
Beilstein 24, 4
Merck Index 15, 3858
Solubility Information Solubility in water: 19g/L (20°C). Other solubilities: 440g/L water (90°C), moderately soluble in methanol, ethanol, ethylene, glycol and pyridine, insoluble in acetone, ether, chloroform, benzene, and ligroin
Formula Weight 102.15
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Imidazolidinethione, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Imidazolidinethione
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360D-May damage the unborn child.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.