missing translation for 'onlineSavingsMsg'
Learn More

2-Imidazolidone, 96%, anhydrous

Product Code. 11314152 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
250 g
1 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
1kg
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11314152 25 g Glass bottle 25g
11976261 250 g Glass bottle 250g
11966261 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11314152 Supplier Thermo Scientific Acros Supplier No. 412270250
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket
This item is not returnable. View return policy

CAS: 120-93-4 | C3H6N2O | 86.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-93-4
Molecular Formula C3H6N2O
Molecular Weight (g/mol) 86.09
MDL Number MFCD00005257
InChI Key YAMHXTCMCPHKLN-UHFFFAOYSA-N
Synonym 2-imidazolidone, 2-imidazolidinone, ethyleneurea, imidazolidinone, 2-oxoimidazolidine, ethylene urea, n,n'-ethyleneurea, 1,3-ethyleneurea, imidazolid-2-one, monoethyleneurea
PubChem CID 8453
ChEBI CHEBI:37001
IUPAC Name imidazolidin-2-one
SMILES O=C1NCCN1

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 131.4°C to 132.4°C
Color White
Boiling Point 187.0°C (10.0 Torr)
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Beilstein 24,2
Merck Index 15,4955
Formula Weight 86.09
Percent Purity 96%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 2-Imidazolidone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.