missing translation for 'onlineSavingsMsg'
Learn More

2-Imidazolidone hemihydrate, 99+%, Thermo Scientific Chemicals

Product Code. 10616512 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
10 kg
Unit Size:
10kg
2.5kg
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10616512 2.5 kg 2.5kg
10182870 25 g 25g
10132540 500 g 500g
10341294 10 kg 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10616512 Supplier Thermo Scientific Acros Supplier No. 122040025
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 121325-67-5 | C6H14N4O3 | 190.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121325-67-5 , 120-93-4
Molecular Formula C6H14N4O3
Molecular Weight (g/mol) 190.20
MDL Number MFCD02662355
InChI Key KPRJGGOOWATRNT-UHFFFAOYSA-N
Synonym imidazolidin-2-one hydrate, 2-imidazolidinone hemihydrate, 2-imidazolidone hemihydrate, bis imidazolidin-2-one hydrate, bis imidazolidinone hydrate, 2-imidazolidinone, hydrate 2:1, imidazolidin-2-one, imidazolidin-2-one, hydrate
PubChem CID 2723651
IUPAC Name imidazolidin-2-one;hydrate
SMILES O.O=C1NCCN1.O=C1NCCN1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 58.0°C
Color White to Yellow
Density 265g/mL
Boiling Point >100.0°C
Flash Point 265°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic bottle
Quantity 2.5 kg
Merck Index 14, 4914
Solubility Information Solubility in water: 950g/L (20°C). Other solubilities: 1800g/L water (40°C)
Formula Weight 95.1
Percent Purity 99+%
Physical Form Adhering Crystals or Crystalline Powder
Chemical Name or Material 2-Imidazolidone hemihydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.