missing translation for 'onlineSavingsMsg'
Learn More

2-Iminothiolane hydrochloride

Product Code. 15422959
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15422959 1 g 1g
15432959 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15422959 Supplier Thermo Scientific Alfa Aesar Supplier No. J60131.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Protein modification and cross-linking reagent, cleavable by thiols | CAS: 4781-83-3 | C4H8ClNS | 137.625 g/mol

2-Iminothiolane hydrochloride is predominantly used as an amine reactive at pH 7-10 and is also an effective thiolation reagent for polysaccharides. It is also used as a RNA-protein crosslinking reagent as well as useful in cross-linking proteins..

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Iminothiolane hydrochloride is predominantly used as an amine reactive at pH 7-10 and is also an effective thiolation reagent for polysaccharides. It is also used as a RNA-protein crosslinking reagent as well as useful in cross-linking proteins..

Solubility
Freely soluble in water

Notes
Protect from moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away fom strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4781-83-3
Molecular Formula C4H8ClNS
Molecular Weight (g/mol) 137.625
MDL Number MFCD00039013
InChI Key ATGUDZODTABURZ-UHFFFAOYSA-N
Synonym 2-iminothiolane hydrochloride, 2-iminothiolane hcl, thiolan-2-imine hydrochloride, dihydrothiophen-2 3h-imine hydrochloride, 2 3h-thiophenimine, dihydro-, hydrochloride, traut's reagent, c4h7ns.clh, traut eo 1/4 a, 2-thiolanimine hydrochloride, 2 3h-thiophenimine, hydrochloride
PubChem CID 13166855
IUPAC Name thiolan-2-imine;hydrochloride
SMILES C1CC(=N)SC1.Cl

Specifications

Melting Point 190°C to 195°C
Color White
Quantity 1 g
Beilstein 3620079
Solubility Information Freely soluble in water
Formula Weight 137.63
Physical Form Powder
Chemical Name or Material 2-Iminothiolane hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.