missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-1,1,1-trifluoroethane, 99%

Product Code. 10174240 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
Unit Size:
10mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10174240 10 mL Glass bottle 10mL
13448418 50 mL Glass bottle 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10174240 Supplier Thermo Scientific Acros Supplier No. 176080100
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 353-83-3 | C2H2F3I | 209.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 353-83-3
Molecular Formula C2H2F3I
Molecular Weight (g/mol) 209.94
MDL Number MFCD00001082
InChI Key RKOUFQLNMRAACI-UHFFFAOYSA-N
Synonym 2-iodo-1,1,1-trifluoroethane, 2,2,2-trifluoroethyl iodide, trifluoroethyl iodide, ethane, 1,1,1-trifluoro-2-iodo, 1-iodo-2,2,2-trifluoroethane, 2,2,2-trifluoroiodoethane, cf3ch2i, unii-ca0j38we9z, 1,1,1-trifluoroiodoethane, 1,1,1-trifluoro-2-iodo-ethane
PubChem CID 67709
IUPAC Name 1,1,1-trifluoro-2-iodoethane
SMILES C(C(F)(F)F)I

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Pink
Density 2.1300g/mL
Boiling Point 53.0°C to 55.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula ICH2CF3
Refractive Index 1.4000 to 1.4020
Quantity 10 mL
Specific Gravity 2.13
Solubility Information Solubility in water: insoluble
Formula Weight 209.94
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Iodo-1, 1, 1-trifluoroethane, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Iodo-1,1,1-trifluoroethane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.