missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-1-(2-tetrahydropyranyl)-1H-imidazole, 95%

Product Code. 15477915
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15477915 250 mg 250mg
15424497 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15477915 Supplier Thermo Scientific Alfa Aesar Supplier No. H31846.MD
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1047626-77-6 | C8H11IN2O | 278.093 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1047626-77-6
Molecular Formula C8H11IN2O
Molecular Weight (g/mol) 278.093
MDL Number MFCD11100955
InChI Key UWFGOSIGMHYERY-UHFFFAOYSA-N
Synonym 2-iodo-1-tetrahydro-2h-pyran-2-yl-1h-imidazole, 2-iodo-1-2-tetrahydropyranyl-1h-imidazole, 2-iodo-1-oxan-2-yl imidazole, 2-iodo-1-oxan-2-yl-1h-imidazole
PubChem CID 45588154
IUPAC Name 2-iodo-1-(oxan-2-yl)imidazole
SMILES C1CCOC(C1)N2C=CN=C2I

Specifications

Quantity 250 mg
Sensitivity Light sensitive
Formula Weight 278.09
Percent Purity 95%
Chemical Name or Material 2-Iodo-1-(2-tetrahydropyranyl)-1H-imidazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.