missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-2-methylpropane, 95%, stab. with copper powder

Product Code. 11327845
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11327845 100 g 100g
11317845 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11327845 Supplier Thermo Scientific Alfa Aesar Supplier No. L02786.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 558-17-8 | C4H9I | 184.02 g/mol

2-Iodo-2-methylpropane is used as a chemical reagent in organic synthesis. Further, it serves as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Iodo-2-methylpropane is used as a chemical reagent in organic synthesis. Further, it serves as a pharmaceutical intermediate.

Solubility
Miscible with ethanol and ether.

Notes
Light sensitive. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 558-17-8
Molecular Formula C4H9I
Molecular Weight (g/mol) 184.02
MDL Number MFCD00001068
InChI Key ANGGPYSFTXVERY-UHFFFAOYSA-N
Synonym tert-butyl iodide, propane, 2-iodo-2-methyl, trimethyliodomethane, t-butyl iodide, unii-e5hy19g10p, 2-jod-2-methylpropan, sjphavimup@, tert-c4h9i, 2-iodo-2-methyl propane, 2-iodo-2-methyl-propane
PubChem CID 11206
IUPAC Name 2-iodo-2-methylpropane
SMILES CC(C)(C)I

Specifications

Melting Point -38°C
Density 1.547
Boiling Point 99°C to 100°C
Flash Point 7°C (44°F)
Odor Recognizable
Refractive Index 1.491
Quantity 100 g
UN Number UN2391
Beilstein 505950
Sensitivity Light sensitive
Solubility Information Miscible with ethanol and ether.
Formula Weight 184.02
Percent Purity 95%
Chemical Name or Material 2-Iodo-2-methylpropane, stabilized with copper powder
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.