missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-5-(trifluoromethyl)pyridine, 96%

Product Code. 15480769
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15480769 5 g 5g
15490769 250 mg 250mg
15470769 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15480769 Supplier Thermo Scientific Alfa Aesar Supplier No. H63848.06
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 100366-75-4 | C6H3F3IN | 273.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100366-75-4
Molecular Formula C6H3F3IN
Molecular Weight (g/mol) 273.00
MDL Number MFCD08741351
InChI Key WSEVWIIOEQZEOU-UHFFFAOYSA-N
PubChem CID 22147841
IUPAC Name 2-iodo-5-(trifluoromethyl)pyridine
SMILES FC(F)(F)C1=CN=C(I)C=C1

Specifications

Quantity 5 g
Sensitivity Light sensitive
Synonym 2-iodo-5-trifluoromethyl pyridine, 5-trifluoromethyl-2-iodopyridine, 2-iodo-5-trifluoromethyl-pyridine, pyridine, 2-iodo-5-trifluoromethyl, pubchem16308, acmc-1c5wh, 2-iodanyl-5-trifluoromethyl pyridine, pyridine,2-iodo-5-trifluoromethyl
Formula Weight 272.99
Percent Purity 96%
Chemical Name or Material 2-Iodo-5-(trifluoromethyl)pyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.