missing translation for 'onlineSavingsMsg'
Learn More

2-Iodoacetamide, TRC

Product Code. 30007345
Change view
Click to view available options
Quantity:
2.5 g
10 g
25 g
Unit Size:
10g
2.5g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007345 2.5 g 2.5g
30006153 10 g 10g
30012354 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007345 Supplier Toronto Research Chemicals Supplier No. TRCI6855052.5G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144-48-9
Molecular Formula C2H4INO
Molecular Weight (g/mol) 184.96
Synonym 2-Iodoacetamide (ACI), Acetamide, iodo- (6CI), Deltop, Iodoacetamide, Monoiodoacetamide, NSC 9581, Surauto, α-Iodoacetamide, Deltop, Iodoacetamide, Monoiodoacetamide, NSC 9581, Surauto, alpha-Iodoacetamide
IUPAC Name 2-iodoacetamide
SMILES NC(=O)CI

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C2H4INO/c3-1-2(4)5/h1H2,(H2,4,5)
Formula Weight 184.9338
Chemical Name or Material 2-Iodoacetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.