missing translation for 'onlineSavingsMsg'
Learn More

2-Iodoethanol, 99%, stab. with copper

Product Code. 11457626
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11457626 25 g 25g
11447626 5 g 5g
11467626 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 11457626 Proveedor Thermo Scientific Alfa Aesar N.º de proveedor A14658.14
 Restricted Item
In Stock
Añadir a la cesta
Añadir a la cesta
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 624-76-0 | C2H5IO | 171.97 g/mol

2-Iodoethanol is used in the preparation of the neuroexcitants such as 4-dicarboxylic acid, alfa-kainic acid, 3-(carboxymethyl)pyrrolidine-2 and alfa-isokainic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Iodoethanol is used in the preparation of the neuroexcitants such as 4-dicarboxylic acid, alfa-kainic acid, 3-(carboxymethyl)pyrrolidine-2 and alfa-isokainic acid.

Solubility
Soluble in water.

Notes
Light sensitive. Store in a cool place. Incompatible with acid anhydrides, acid chlorides, phosphorus halides, strong acids, strong oxidizing agents and strong reducing agents.
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 624-76-0
Molecular Formula C2H5IO
Molecular Weight (g/mol) 171.97
MDL Number MFCD00002830
InChI Key QSECPQCFCWVBKM-UHFFFAOYSA-N
Synonym ethanol, 2-iodo, iodoethanol, ethylene iodohydrin, 2-iodoethan-1-ol, iodoethyl alcohol, 2-indoethanol, 2-iodo ethanol, 2-iodo-ethanol, 2-iodo-1-ethanol, 2-iodoethyl alcohol
PubChem CID 12225
IUPAC Name 2-iodoethanol
SMILES OCCI

Especificaciones

Density 2.202
Boiling Point 85°C (25 mmHg)
Flash Point 65°C (149°F)
Odor Characteristic
Linear Formula ICH2CH2OH
Refractive Index 1.572
Quantity 25 g
UN Number UN2810
Beilstein 1098258
Sensitivity Light sensitive
Solubility Information Soluble in water.
Formula Weight 171.96
Percent Purity 99%
Chemical Name or Material 2-Iodoethanol, stabilized with copper
Mostrar más Mostrar menos
compliance-icons
Nombre del producto
  • 2-Iodoethanol
Palabra de advertencia
  • Danger
Clasificación
  • Acute toxicity Category 2
Indicaciones de peligro
  • H300-Fatal if swallowed.
Consejos de prudencia
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P330-Rinse mouth.
Otros peligros
  • MIXTURE LIST-Contain: 2-Iodoethanol

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.