missing translation for 'onlineSavingsMsg'
Learn More

2-Iodoethanol, 99%, stab. with copper

Product Code. 11467626
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11467626 100 g 100g
11447626 5 g 5g
11457626 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11467626 Supplier Thermo Scientific Alfa Aesar Supplier No. A14658.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 624-76-0 | C2H5IO | 171.97 g/mol

2-Iodoethanol is used in the preparation of the neuroexcitants such as 4-dicarboxylic acid, alfa-kainic acid, 3-(carboxymethyl)pyrrolidine-2 and alfa-isokainic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Iodoethanol is used in the preparation of the neuroexcitants such as 4-dicarboxylic acid, alfa-kainic acid, 3-(carboxymethyl)pyrrolidine-2 and alfa-isokainic acid.

Solubility
Soluble in water.

Notes
Light sensitive. Store in a cool place. Incompatible with acid anhydrides, acid chlorides, phosphorus halides, strong acids, strong oxidizing agents and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 624-76-0
Molecular Formula C2H5IO
Molecular Weight (g/mol) 171.97
MDL Number MFCD00002830
InChI Key QSECPQCFCWVBKM-UHFFFAOYSA-N
Synonym ethanol, 2-iodo, iodoethanol, ethylene iodohydrin, 2-iodoethan-1-ol, iodoethyl alcohol, 2-indoethanol, 2-iodo ethanol, 2-iodo-ethanol, 2-iodo-1-ethanol, 2-iodoethyl alcohol
PubChem CID 12225
IUPAC Name 2-iodoethanol
SMILES OCCI

Specifications

Density 2.202
Boiling Point 85°C (25 mmHg)
Flash Point 65°C (149°F)
Odor Characteristic
Linear Formula ICH2CH2OH
Refractive Index 1.572
Quantity 100 g
UN Number UN2810
Beilstein 1098258
Sensitivity Light sensitive
Solubility Information Soluble in water.
Formula Weight 171.96
Percent Purity 99%
Chemical Name or Material 2-Iodoethanol, stabilized with copper
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.