missing translation for 'onlineSavingsMsg'
Learn More

2-Iodoethanol, 99%, stabilized

Product Code. 10551431 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
250 mL
Unit Size:
10mL
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10551431 10 mL 10mL
10561431 50 mL 50mL
11914641 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10551431 Supplier Thermo Scientific Acros Supplier No. 175500100
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 624-76-0 | C2H5IO | 171.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 624-76-0
Molecular Formula C2H5IO
Molecular Weight (g/mol) 171.97
MDL Number MFCD00002830
InChI Key QSECPQCFCWVBKM-UHFFFAOYSA-N
Synonym ethanol, 2-iodo, iodoethanol, ethylene iodohydrin, 2-iodoethan-1-ol, iodoethyl alcohol, 2-indoethanol, 2-iodo ethanol, 2-iodo-ethanol, 2-iodo-1-ethanol, 2-iodoethyl alcohol
PubChem CID 12225
SMILES OCCI

Specifications

Density 2.2050g/mL
Boiling Point 172.0°C to 175.0°C
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula ICH2CH2OH
Refractive Index 1.5690 to 1.5720
Quantity 10 mL
Beilstein 01,339
Fieser 13,348
Specific Gravity 2.205
IUPAC Name 2-iodoethanol
Stabilizer Copper
Formula Weight 171.97
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Iodoethanol
Show More Show Less
compliance-icons
Product Identifier
  • 2-Iodoethanol, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
Hazard Statement
  • H300-Fatal if swallowed.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P330-Rinse mouth.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.