missing translation for 'onlineSavingsMsg'
Learn More

2-Iodoethanol, 99%, stabilized

Product Code. 11914641 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
250 mL
Unit Size:
10mL
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11914641 250 mL 250mL
10551431 10 mL 10mL
10561431 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11914641 Supplier Thermo Scientific Acros Supplier No. 175502500
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 624-76-0 | C2H5IO | 171.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 624-76-0
Molecular Formula C2H5IO
Molecular Weight (g/mol) 171.97
MDL Number MFCD00002830
InChI Key QSECPQCFCWVBKM-UHFFFAOYSA-N
Synonym ethanol, 2-iodo, iodoethanol, ethylene iodohydrin, 2-iodoethan-1-ol, iodoethyl alcohol, 2-indoethanol, 2-iodo ethanol, 2-iodo-ethanol, 2-iodo-1-ethanol, 2-iodoethyl alcohol
PubChem CID 12225
SMILES OCCI

Tekniske data

Density 2.2050g/mL
Boiling Point 172.0°C to 175.0°C
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Linear Formula ICH2CH2OH
Refractive Index 1.5690 to 1.5720
Quantity 250 mL
Beilstein 01,339
Fieser 13,348
Specific Gravity 2.205
IUPAC Name 2-iodoethan-1-ol
Stabilizer Copper
Formula Weight 171.97
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Iodoethanol
Vis mere Vis mindre
compliance-icons
Produktidentifikator
  • 2-Iodoethanol, stabilized
Signalord
  • Danger
Farekategori
  • Acute toxicity Category 2
Faresætning
  • H300-Fatal if swallowed.
Sikkerhedssætning
  • P264-Wash face, hands and any thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P330-Rinse mouth.

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.